missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Flash column CHROMABOND Flash length 154mm x 26.8mm I.D. FM 70/10 adsorbent weight 10 g

Brand:  Macherey Nagel 730768

Flash column CHROMABOND Flash length 154mm x 26.8mm I.D. FM 70/10 adsorbent weight 10 g

Code : 50

Additional Details : Weight : 0.50000kg

Product Code. 12831031

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.