missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chest freezer ext dims 2225mm x 859mm x 1029mm (w x d x h) 230V 50Hz 481L capacity

Brand:  Thermo Scientific I917CV

Chest freezer ext dims 2225mm x 859mm x 1029mm (w x d x h) 230V 50Hz 481L capacity

Code : 46

Additional Details : Weight : 372.00000kg

Product Code. 12616035

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.