missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 123count eBeads™ Counting Beads

Product Code. 15526296
Change view
Click to view available options
Quantity:
100 tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15526296 100 tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains Sodium azide
Product Code. 15526296 Supplier Invitrogen™ Supplier No. 01123442
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Contains Sodium azide

Intended for use in absolute counting of cells or other particles by flow cytometry. 123count eBeads are 7μm microparticles containing encapsulated dyes compatible with blue (488nm) and violet (405nm) excitation sources and emitting fluorescence between approximately 500nm and 750nm.

  • Supplied at a known concentration in 0.05% Tween-20 with 2 mM sodium azide
  • 10mL aqueous suspension
  • Collecting carefully suspended and measured quantities of cell:bead mixtures on a flow cytometer makes the ratiometric enumeration of cells possible
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Flow Cytometer
No. of Tests 100 tests
Quantity 100 tests
Product Line eBeads
Product Type Cell Counting Bead
I am using counting beads to count cells, but I cannot find the beads on my scatter plots. What do I do?

The first thing to do is check your threshold and see if it is set on forward scatter. If so, the beads are probably being excluded by the threshold. Reducing the threshold setting should reveal your beads.

I want to count my cells using flow cytometry. How do I do this?

Cell counting using flow cytometry can be accomplished by adding an internal microsphere counting standard to the flow cytometric sample. The number of reference beads that are collected reflects a known volume. This allows you to calculate cell concentration.

What does yield and purity mean in reference to the MagniSort kits? How are each of these parameters calculated?

Yield is the percentage of target cells harvested based on the number of cells theoretically available in the starting population. To calculate yield, stain a sample of unsorted and sorted cells with antibodies to identify the target cells, and count the total number of target cells using 123count eBeads (Cat. No. 01-1234-42) on the cytometer. Yield = # of sorted target cells, # of target cells unsorted.

Purity is the percentage of cells harvested that are target cells. Purity can be directly generated by flow cytometric analysis of sorted samples. To obtain purity, stain the sorted cells with antibodies to identify the target cells and analyze by flow cytometry. Gate on live, single cells, and subsequently gate on the target cell population. Purity is the percentage of target cells in the live, single-cell gate.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.