missing translation for 'onlineSavingsMsg'
Learn More

Helios Italquartz 1234 DIN Standard Covers with Ring Handle, Transparent

Product Code. 15888084
Click to view available options
For Use With (Equipment):
CR 41-61 Crucible
CR 42-51-62 Crucible
CR 43-52-63 Crucible
CR 44-53-64 Crucible
CR 45-54-65 Crucible
CR 46-55-66 Crucible
CR 47-56 Crucible
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15888084

Brand: Helios Italquartz 64P00601

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

1234 DIN standard covers with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.

  • Transparent
  • Include ring handle
  • Conform to 1234 DIN standard
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass

Specifications

Color Transparent
Material Quartz
Type 1234 DIN Standard Covers with Ring Handle
For Use With (Equipment) CR 41-61 Crucible
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.