missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMS™ Series 4 Model 1200A Cooled Incubator, 1200 L, Stainless Steel
Description
- Reliable and accurate temperature control
- Temperature range -10.0°C to +50.0°C
- Models within this series include 600L and 1200L capacity incubators as either single or dual temperature (24 hour or 7 day cycle) versions, or a fully programmable 1200L capacity incubator providing multiple temperature control
- Stainless steel exterior and interior construction with foamed polyurethane insulation
- Energy efficient and eco-friendly
- P.I.D. heating and cooling to ensure stable internal temperature
- Pre-set safety cut-out
- Hermetically sealed refrigeration system
- Door lock
- Controls easy to read and simple to use
- Temperature variation with average workload ±0.5°C
- Full range of factory-fitted optional extras available, including LED lighting, access porthole, inner perspex doors, outer glazed doors, alarm system, internal socket and chart recorder
- Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008 |
| Controller Type | Standard digital with Automatic Dual Temperature Cycling |
| Description | Freestanding incubator |
| Capacity (Metric) | 1200 L |
| Chamber Material | Stainless Steel |
| Convection Type | Fan-assisted Circulation |
| Dimensions (D x W) Shelf | 550 mm x 530 mm |
| Lighting | Optional LED |
| Material | Stainless Steel |
| No. of Shelves | 10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
