Learn More
12-Tungstophosphate hydrate, Reagent Grade, Thermo Scientific Chemicals
£59.50 - £177.00
Chemical Identifiers
CAS | 12501-23-4 |
---|---|
Molecular Formula | H5O41PW12 |
Molecular Weight (g/mol) | 2880.20 |
MDL Number | MFCD00011342 |
InChI Key | AVFBYUADVDVJQL-UHFFFAOYSA-N |
Synonym | phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade |
PubChem CID | 16212977 |
SMILES | O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361868
|
Thermo Scientific Alfa Aesar
011827.18 |
50 g |
£59.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351868
|
Thermo Scientific Alfa Aesar
011827.30 |
250 g |
£177.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
12-Tungstophosphate hydrate is used in histology as a component for staining of cell specimens. It binds to fibrin, collagen and fibers of connective tissues. It is also used to replace the anions of dyes from these materials. In solution, it acts as a homogeneous catalyst and involved in the hydrolysis of propene to give 2-propanol. It is supported with alumina and silica and act as a heterogeneous catalyst involved in the dehydration of 2-propanol to propene and methanol to hydrocarbons. In analytical chemistry, it is used to precipitate proteins and termed as universal precipitant for polar proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12501-23-4 | |
2880.20 | |
AVFBYUADVDVJQL-UHFFFAOYSA-N | |
16212977 |
H5O41PW12 | |
MFCD00011342 | |
phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade | |
O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O |
Specifications
95°C | |
White through Yellow | |
H5O41PW12 | |
UN3260 | |
Soluble in water,dichloromethane,acetonitrile,acetic acid and ionic liquids. | |
O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O | |
2880.20 | |
2880.20 (Anhydrous) | |
Moisture & Light Sensitive |
12501-23-4 | |
Powder | |
MFCD00011342 | |
phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade | |
AVFBYUADVDVJQL-UHFFFAOYSA-N | |
phosphoric acid dodecakis(trioxotungsten) hydrate | |
16212977 | |
Reagent | |
12-Tungstophosphate hydrate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only