missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 12-Place Swinging Bucket Microliter Rotor with Cover

For use with Sorvall Primo and Primo R Centrifuges.

Brand:  Thermo Scientific™ 75007592

Product Code. 10464472

  • £1360.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High RCF of 16,438 x g
  • Separation of liquid multi-phase systems
  • Centrifugation of filtration units
  • Density gradient-separation of cells/cell organelles
  • Semi-preparative centrifugation of proteins
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Swinging Bucket Microliter Rotor
16,438 x g
Thermo Scientific Sorvall Primo R Centrifuge
12 x 1.5/2 mL
13,000 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers