missing translation for 'onlineSavingsMsg'
Learn More

12-Molybdophosphoric acid hydrate, Mo >47.5%

Product Code. 11302797
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302797 100 g 100g
11392787 25 g 25g
11312797 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302797 Supplier Thermo Scientific Alfa Aesar Supplier No. L15827.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol

12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

Solubility
Soluble in water, ethanol and ether.

Notes
Hygroscopic. Incompatible with organic materials and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51429-74-4
Molecular Formula H3Mo12O40P
Molecular Weight (g/mol) 1825.40
MDL Number MFCD00149913
InChI Key FEFSFHGZSNHJEL-UHFFFAOYSA-N
Synonym Molybdophosphoric acid
IUPAC Name phosphoric acid dodecamolybdenum hexatriacontaoxidandiide
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O

Specifications

Melting Point 78°C to 90°C
Density 3.15
Sensitivity Hygroscopic
Quantity 100 g
UN Number UN3085
Merck Index 14,7342
Solubility Information Soluble in water,ethanol and ether.
Formula Weight 1825.25 (Anhydrous)
Assay Mo >47.5%
Chemical Name or Material 12-Molybdophosphoric acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.