missing translation for 'onlineSavingsMsg'
Learn More

12-Mercaptododecylphosphonic acid, 95%

Product Code. 15456867
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456867 250 mg 250mg
15446867 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456867 Supplier Thermo Scientific Alfa Aesar Supplier No. H33969.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 159239-33-5 | C12H27O3PS | 282.38 g/mol

Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159239-33-5
Molecular Formula C12H27O3PS
Molecular Weight (g/mol) 282.38
MDL Number MFCD11046116
InChI Key PVIUMTORLYKRJT-UHFFFAOYSA-N
Synonym 12-mercaptododecylphosphonic acid, 12-sulfanyldodecyl phosphonic acid
PubChem CID 45158994
IUPAC Name 12-sulfanyldodecylphosphonic acid
SMILES OP(O)(=O)CCCCCCCCCCCCS

Specifications

Melting Point 94°C to 96°C
Linear Formula HS(CH2)12P(O)(OH)2
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 282.38
Percent Purity 95%
Chemical Name or Material 12-Mercaptododecylphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.