missing translation for 'onlineSavingsMsg'
Learn More
Learn More
12-Lipoxygenase Antibody (SR2151), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22243-100ul
This item is not returnable.
View return policy
Description
12-Lipoxygenase Monoclonal antibody specifically detects 12-Lipoxygenase in Human, Rat samples. It is validated for Western Blot
Specifications
| 12-Lipoxygenase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| SR2151 | |
| Western Blot 1:500-1:2000 | |
| 12(S)-lipoxygenase, 12-LOX, 12S-lipoxygenase, 12S-LOXarachidonate 12-lipoxygenase, 12S-type, arachidonate 12-lipoxygenase, EC 1.13.11, EC 1.13.11.31, LOG12, platelet 12-LOX, platelet-type 12-lipoxygenase, Platelet-type lipoxygenase 12 | |
| A synthesized peptide derived from human 12-Lipoxygenase (Uniprot #: P18054) | |
| 100 μg | |
| Lipid and Metabolism | |
| 239 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction