missing translation for 'onlineSavingsMsg'
Learn More
Learn More
12-Lipoxygenase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35458-20ul
This item is not returnable.
View return policy
Description
12-Lipoxygenase Polyclonal antibody specifically detects 12-Lipoxygenase in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunofluorescence
Specifications
| 12-Lipoxygenase | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunofluorescence 1:50 - 1:200 | |
| 12(S)-lipoxygenase, 12-LOX, 12S-lipoxygenase, 12S-LOXarachidonate 12-lipoxygenase, 12S-type, arachidonate 12-lipoxygenase, EC 1.13.11, EC 1.13.11.31, LOG12, platelet 12-LOX, platelet-type 12-lipoxygenase, Platelet-type lipoxygenase 12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 483-663 of human 12-Lipoxygenase (NP_000688.2).,, Sequence:, YQRDDIVKGDPELQAWCREITEVGLCQAQDRGFPVSFQSQSQLCHFLTMCVFTCTAQHAAINQGQLDWYAWVPNAPCTMRMPPPTTKEDVTMATVMGSLPDVRQACLQMAISWHLSRRQPDMVPLGHHKEKYFSGPKPKAVLNQFRTDLEKLEKEITARNEQLDWPYEYLKPSCIENSVTI | |
| 20 μL | |
| Lipid and Metabolism | |
| 239 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction