missing translation for 'onlineSavingsMsg'
Learn More

12-Ketostearic Acid Sodium Salt, TRC

Product Code. 30402082
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402082 100 mg
30408004 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402082 Supplier Toronto Research Chemicals Supplier No. TRCK264901100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73536-57-9
Molecular Formula C18H33NaO3
Molecular Weight (g/mol) 320.44
Synonym 12-Oxooctadecanoic Acid Sodium Salt, 12-Ketooctadecanoic Acid Sodium Salt, 12-Oxostearic Acid Sodium Salt, Lycaonic Acid Sodium Salt, NSC 2103
IUPAC Name sodium;12-oxooctadecanoate
SMILES Nc1ccc(cc1)N2C(=O)COCC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H34O3.Na/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21;/h2-16H2,1H3,(H,20,21);/q;+1/p-1
Formula Weight 320.2327
Chemical Name or Material 12-Ketostearic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.