missing translation for 'onlineSavingsMsg'
Learn More

12-Hydroxystearic Acid, TRC

Product Code. 30403747
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403747 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403747 Supplier Toronto Research Chemicals Supplier No. TRCH9537802.5G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 12-Hydroxystearic Acid
CAS 106-14-9
Molecular Formula C18 H36 O3
Quantity 2.5 g
Synonym Octadecanoic acid, 12-hydroxy-, Stearic acid, λ-hydroxy- (4CI), 12-Hydroxyoctadecanoic acid, 12-HSA, 12-Hydroacid, 12-Hydroacid HP, 12-Hydroxystearic acid, 12HAS-I, ADK Stab LS 12, Barolub FTO, Cerit Fac 3, Ceroxin GL, DL-12-Hydroxyoctadecanoic acid, DL-12-Hydroxystearic acid, G 21, H 0308, HSA, HSIM 160, Harwax A, Hoctol A, Hydrofol Acid 200, Hydroxystearin R, KOW, Loxiol G 21, NSC 2385, Pelemol HSA, Solid X, Solsperse 2000
InChI Formula InChI=1S/C18H36O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h17,19H,2-16H2,1H3,(H,20,21)
SMILES CCCCCCC(O)CCCCCCCCCCC(=O)O
IUPAC Name 12-hydroxyoctadecanoic acid
Molecular Weight (g/mol) 300.48
Formula Weight 300.2664
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.