missing translation for 'onlineSavingsMsg'
Learn More

12-Hydroxystearic Acid Lithium Salt, TRC

Product Code. 30398169
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398169 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398169 Supplier Toronto Research Chemicals Supplier No. TRCH95378510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium 12-Hydroxystearate
CAS 7620-77-1
Molecular Formula C18 H35 O3 . Li
Quantity 10 g
Synonym 12-Hydroxystearic Acid Lithium Salt, Octadecanoic acid, 12-hydroxy-, lithium salt (1:1), Octadecanoic acid, 12-hydroxy-, monolithium salt (8CI,9CI), 12-Hydroxystearic acid lithium salt, Daiwax OHL, LS 6, Lithium 12-hydroxyoctadecanoate, Lithium 12-hydroxystearate, NLGI 3, OHL, S 7000H, S 700H
InChI Formula InChI=1S/C18H36O3.Li/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21;/h17,19H,2-16H2,1H3,(H,20,21);/q;+1/p-1
SMILES [Li+].CCCCCCC(O)CCCCCCCCCCC(=O)[O-]
IUPAC Name lithium;12-hydroxyoctadecanoate
Molecular Weight (g/mol) 306.41
Formula Weight 306.275
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.