missing translation for 'onlineSavingsMsg'
Learn More
Learn More
12-Hydroxystearic Acid Lithium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Lithium 12-Hydroxystearate |
| CAS | 7620-77-1 |
| Molecular Formula | C18 H35 O3 . Li |
| Quantity | 1 g |
| Synonym | 12-Hydroxystearic Acid Lithium Salt, Octadecanoic acid, 12-hydroxy-, lithium salt (1:1), Octadecanoic acid, 12-hydroxy-, monolithium salt (8CI,9CI), 12-Hydroxystearic acid lithium salt, Daiwax OHL, LS 6, Lithium 12-hydroxyoctadecanoate, Lithium 12-hydroxystearate, NLGI 3, OHL, S 7000H, S 700H |
| InChI Formula | InChI=1S/C18H36O3.Li/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21;/h17,19H,2-16H2,1H3,(H,20,21);/q;+1/p-1 |
| SMILES | [Li+].CCCCCCC(O)CCCCCCCCCCC(=O)[O-] |
| IUPAC Name | lithium;12-hydroxyoctadecanoate |
| Molecular Weight (g/mol) | 306.41 |
| Formula Weight | 306.275 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?