missing translation for 'onlineSavingsMsg'
Learn More
Learn More
12-Hydroxystearic Acid-d5, TRC
Specifications
Specifications
| Molecular Formula | C18 D5 H31 O3 |
| Quantity | 2.5 mg |
| Synonym | 12-Hydroxyoctadecanoic Acid-d5, lambda-Hydroxystearic Acid-d5, 12-HSA-d5, ADK Stab LS 12-d5, Barolub FTO-d5, Cerit Fac 3-d5, Ceroxin GL-d5, DL-12-Hydroxyoctadecanoic Acid-d5, DL-12-Hydroxystearic Acid-d5, G 21-d5, H 0308-d5, HSIM 160-d5, Harwax A-d5, Hoctol A-d5, Hydrofol Acid 200-d5, Hydroxystearin R-d5, KOW-d5, Loxiol G 21-d5, NSC 2385-d5, Pelemol HSA-d5, Solid X-d5, Solsperse 2000-d5 |
| InChI Formula | InChI=1S/C18H36O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h17,19H,2-16H2,1H3,(H,20,21)/i14D2,15D2,17D |
| SMILES | [2H]C([2H])(CCCCC)C([2H])(O)C([2H])([2H])CCCCCCCCCC(=O)O |
| IUPAC Name | 11,11,12,13,13-pentadeuterio-12-hydroxyoctadecanoic acid |
| Molecular Weight (g/mol) | 305.507 |
| Formula Weight | 305.298 |
| Chemical Name or Material | 12-Hydroxystearic Acid-d5 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?