missing translation for 'onlineSavingsMsg'
Learn More

12-Hydroxy Irinotecan (Mixture of Diastereomers), TRC

Product Code. 30395701
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395701 10 mg
30392331 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395701 Supplier Toronto Research Chemicals Supplier No. TRCH94354510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185336-12-3
Molecular Formula C33 H38 N4 O7
Molecular Weight (g/mol) 602.68
Synonym [1,4'-Bipiperidine]-1'-carboxylic acid, (4S)-4,11-diethyl-3,4,12,14-tetrahydro-4,12-dihydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl ester, [1,4'-Bipiperidine]-1'-carboxylic acid, 4,11-diethyl-3,4,12,14-tetrahydro-4,12-dihydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl ester, (4S)-, RPR 112524, (4S)-4,11-Diethyl-4,12-dihydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl 1,4'-bipiperidine-1'-carboxylate, 12-Hydroxy Irinotecan
SMILES CCc1c2C(O)N3C(=O)C4=C(C=C3c2nc5ccc(OC(=O)N6CCC(CC6)N7CCCCC7)cc15)[C@@](O)(CC)C(=O)OC4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C33H38N4O7/c1-3-21-22-16-20(44-32(41)36-14-10-19(11-15-36)35-12-6-5-7-13-35)8-9-25(22)34-28-26-17-24-23(18-43-31(40)33(24,42)4-2)29(38)37(26)30(39)27(21)28/h8-9,16-17,19,30,39,42H,3-7,10-15,18H2,1-2H3/t30?,33-/m0/s1
Formula Weight 602.274
Chemical Name or Material 12-hydroxy Irinotecan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.