Learn More
12-Crown-4, 98%
Description
It serves as an aprotic solvent for alkali metals, the sodium solution is a strong reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It serves as an aprotic ″solvent″ for alkali metals, the sodium solution is a strong reducing agent.
Solubility
Fully miscible in water.
Notes
Light & Air Sensitive & Hygroscopic. Store under inert gas. Keep in a tightly closed container.
Specifications
Specifications
| Melting Point | 17°C |
| Density | 1.108 |
| Boiling Point | 68°C to 70°C (0.1 mmHg) |
| Flash Point | >109°C (228°F) |
| Odor | Odorless |
| Refractive Index | 1.462 |
| Quantity | 5 g |
| UN Number | UN2810 |
| Beilstein | 1363064 |
| Sensitivity | Air and light sensitive; Hygroscopic |
| Show More |
Safety and Handling
- 12-Crown-4
- Danger
- Acute toxicity Category 1
- H330-Fatal if inhaled.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: 12-Crown-4
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.