missing translation for 'onlineSavingsMsg'
Learn More

11Beta-Hydroxy Dienogest, TRC

Product Code. 30393812
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393812 10 mg -
30390141 1 mg 1mg
30407366 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393812 Supplier Toronto Research Chemicals Supplier No. TRCH90540010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86153-39-1
Molecular Formula C20 H25 N O3
Molecular Weight (g/mol) 327.42
Synonym 19-Norpregna-4,9-diene-21-nitrile, 11,17-dihydroxy-3-oxo-, (11β,17α)- (9CI), 11β-Hydroxy-STS 557, 17α-Cyanomethyl-11β,17β-dihydroxyestra-4,9-dien-3-one, STS 749, 11β-Hydroxydienogest, 11β-Hydroxy dienogest, (11β,17-Dihydroxy-3-oxoestra-4,9-dien-17α-yl)acetonitrile
IUPAC Name 2-[(8S,11S,13S,14S,17R)-11,17-dihydroxy-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]acetonitrile
SMILES C[C@]12C[C@H](O)C3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@]2(O)CC#N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H25NO3/c1-19-11-17(23)18-14-5-3-13(22)10-12(14)2-4-15(18)16(19)6-7-20(19,24)8-9-21/h10,15-17,23-24H,2-8,11H2,1H3/t15-,16-,17-,19-,20+/m0/s1
Formula Weight 327.1834
Chemical Name or Material 11beta-Hydroxydienogest
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.