missing translation for 'onlineSavingsMsg'
Learn More

11Beta-Hydroxy-3-oxo-androsta-1,4-diene-17Beta-carboxylic Acid, TRC

Product Code. 30396659
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396659 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396659 Supplier Toronto Research Chemicals Supplier No. TRCH9502351MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11beta-Hydroxy-3-oxo-androsta-1,4-diene-17beta-carboxylic acid
CAS 75448-51-0
Molecular Formula C20 H26 O4
Quantity 1 mg
Synonym Androsta-1,4-diene-17-carboxylic acid, 11-hydroxy-3-oxo-, (11β,17β)-, Androsta-1,4-diene-17β-carboxylic acid, 11β-hydroxy-3-oxo- (6CI), (11β,17β)-11-Hydroxy-3-oxoandrosta-1,4-diene-17-carboxylic acid, 11β-Hydroxy-3-oxo-androsta-1,4-diene-17β-carboxylic acid
InChI Formula InChI=1S/C20H26O4/c1-19-8-7-12(21)9-11(19)3-4-13-14-5-6-15(18(23)24)20(14,2)10-16(22)17(13)19/h7-9,13-17,22H,3-6,10H2,1-2H3,(H,23,24)/t13-,14-,15+,16-,17+,19-,20-/m0/s1
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)O
IUPAC Name (8S,9S,10R,11S,13S,14S,17S)-11-hydroxy-10,13-dimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-17-carboxylic acid
Molecular Weight (g/mol) 330.42
Formula Weight 330.1831
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.