missing translation for 'onlineSavingsMsg'
Learn More

(11Beta,17Z)-11,20-Dihydroxy-3-oxo-pregna-4,17(20)-dien-21-al, TRC

Product Code. 30372557
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372557 50 mg -
30369007 100 mg -
30374939 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372557 Supplier Toronto Research Chemicals Supplier No. TRCD45391050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105562-13-8
Molecular Formula C21H28O4
Molecular Weight (g/mol) 344.44
Synonym Pregna-4,17(20)-dien-21-al, 11,20-dihydroxy-3-oxo-, (11β,17Z)-, (11β,17Z)-11,20-Dihydroxy-3-oxopregna-4,17(20)-dien-21-al
IUPAC Name (2Z)-2-hydroxy-2-[(8S,9S,10R,11S,13S,14S)-11-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-ylidene]acetaldehyde
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC/C/2=C(/O)\C=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H28O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,11,14-15,17,19,24-25H,3-8,10H2,1-2H3/b18-16-/t14-,15-,17-,19+,20-,21-/m0/s1
Formula Weight 344.1988
Chemical Name or Material (11beta,17Z)-11,20-Dihydroxy-3-oxo-pregna-4,17(20)-dien-21-al
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.