missing translation for 'onlineSavingsMsg'
Learn More

(11Beta,16Beta)-3,21-(Dibenzoyloxy)-9-fluoro-11,17-dihydroxy-16-methyl-pregna-1,3,5-trien-20-one, TRC

Product Code. 30377189
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377189 100 mg 100mg
30380805 250 mg 250mg
30377790 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377189 Supplier Toronto Research Chemicals Supplier No. TRCD422515100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (11beta,16beta)-3,21-(Dibenzoyloxy)-9-fluoro-11,17-dihydroxy-16-methyl-pregna-1,3,5-trien-20-one
Analyte or Component Names (11beta,16beta)-3,21-(Dibenzoyloxy)-9-fluoro-11,17-dihydroxy-16-methyl-pregna-1,3,5-trien-20-one
Shipping Condition Room Temperature
Molecular Formula C36H37FO7
InChI Formula InChI=1S/C36H37FO7/c1-22-18-28-27-15-14-25-19-26(44-32(41)24-12-8-5-9-13-24)16-17-33(25,2)35(27,37)29(38)20-34(28,3)36(22,42)30(39)21-43-31(40)23-10-6-4-7-11-23/h4-14,16-17,19,22,27-29,38,42H,15,18,20-21H2,1-3H3/t22-,27-,28-,29-,33-,34-,35-,36-/m0/s1
Solution Type Neat
SMILES C[C@@H](C1)[C@@](C(COC(C2=CC=CC=C2)=O)=O)(O)[C@]3(C)[C@]1([H])[C@]4([H])CC=C5C=C(OC(C6=CC=CC=C6)=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C3
IUPAC Name 2-((8S,9R,10S,11S,13S,14S,16S,17R)-3-(benzoyloxy)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-8,9,10,11,12,13,14,15,16,17-decahydro-7H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl benzoate
Industry Type Forensics
Molecular Weight (g/mol) 600.67
Quantity 100 mg
Formula Weight 600.25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.