missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(11Beta,16Alpha)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)-pregna-1,4-diene-3,20-dione Monosodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Dexamethasone monosodium phosphate |
| CAS | 1869-92-7 |
| Molecular Formula | C22 H29 F O8 P . Na |
| Quantity | 250 mg |
| Synonym | 9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)-pregna-1,4-diene-3,20-dione Monosodium Salt, 9-Fluoro-11β,17,21-trihydroxy-16α-methyl-pregna-1,4-diene-3,20-dione 21-Phosphate Monosodium Salt, (11Beta,16Alpha)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)-pregna-1,4-diene-3,20-dione Monosodium Salt |
| InChI Formula | InChI=1S/C22H30FO8P.Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);/q;+1/p-1/t12-,15+,16+,17+,19+,20+,21+,22+;/m1./s1 |
| SMILES | [Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)(O)[O-] |
| IUPAC Name | sodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxo-ethyl] hydrogen phosphate |
| Molecular Weight (g/mol) | 494.42 |
| Formula Weight | 494.1482 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?