missing translation for 'onlineSavingsMsg'
Learn More

(11b,17a)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-3-oxo-androsta-1,4-diene-17-carboxylic Acid, TRC

Product Code. 30387131
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387131 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387131 Supplier Toronto Research Chemicals Supplier No. TRCE8905702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 133991-63-6
Molecular Formula C23H30O7
Quantity 2.5 g
Synonym PJ 91, Dechloromethyl Loteprednol Etabonate
InChI Formula InChI=1S/C23H30O7/c1-4-29-20(28)30-23(19(26)27)10-8-16-15-6-5-13-11-14(24)7-9-21(13,2)18(15)17(25)12-22(16,23)3/h7,9,11,15-18,25H,4-6,8,10,12H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,21-,22-,23-/m0/s1
SMILES OC(=O)C1[C@H](O)C(O)C(O)[C@@H](O1)OC2[C@@H](OC(C(O)=O)[C@H](O)C2O)O[C@@H]3C(C)(C)[C@@H]4[C@@](CC3)(C)[C@H]5[C@](CC4)(C)[C@]6(C)C(=CC5=O)[C@H]7[C@@](CCC(C(O)=O)(C)C7)(C)CC6
Molecular Weight (g/mol) 418.48
Formula Weight 418.1992
Chemical Name or Material (11beta,17alpha)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-3-oxo-androsta-1,4-diene-17-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.