missing translation for 'onlineSavingsMsg'
Learn More

11Alpha-Hydroxy Mexrenone, TRC

Product Code. 30396442
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396442 100 mg
30400005 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396442 Supplier Toronto Research Chemicals Supplier No. TRCH948140100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192704-56-6
Molecular Formula C24H32O6
Molecular Weight (g/mol) 416.51
Synonym (7alpha,11alpha,17alpha)-11,17-Dihydroxy-3-oxopregn-4-ene-7,21-dicarboxylic Acid gamma-Lactone Methyl Ester, 11alpha,17beta-Dihydroxypregn-4-en-3-one 7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester, 11alpha-Hydroxymexrenone
IUPAC Name methyl (7R,8S,9S,10R,11R,13S,14S,17R)-11-hydroxy-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-carboxylate
SMILES COC(=O)[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]3[C@H](O)C[C@@]4(C)[C@@H](CC[C@@]45CCC(=O)O5)[C@H]13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H32O6/c1-22-7-4-14(25)10-13(22)11-15(21(28)29-3)19-16-5-8-24(9-6-18(27)30-24)23(16,2)12-17(26)20(19)22/h10,15-17,19-20,26H,4-9,11-12H2,1-3H3/t15-,16+,17-,19+,20+,22+,23+,24-/m1/s1
Formula Weight 416.2199
Chemical Name or Material 11alpha-Hydroxy Mexrenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.