missing translation for 'onlineSavingsMsg'
Learn More

11Alpha,17Alpha,21-Trihydroxy-pregn-5-ene-3,20-dione 3,20-Diethylene Ketal, TRC

Product Code. 30453027
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453027 Supplier Toronto Research Chemicals Supplier No. TRCT79524550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 11alpha,17alpha,21-Trihydroxy-pregn-5-ene-3,20-dione 3,20-Diethylene Ketal
CAS 102030-55-7
Molecular Formula C25H38O7
Quantity 50 mg
Synonym 11,17,21-Trihydroxy-pregn-5-ene-3,20-dione Cyclic 3,20-Bis(1,2-ethanediyl Acetal)
InChI Formula InChI=1S/C25H38O7/c1-21-7-8-23(29-9-10-30-23)13-16(21)3-4-17-18-5-6-24(28,25(15-26)31-11-12-32-25)22(18,2)14-19(27)20(17)21/h3,17-20,26-28H,4-15H2,1-2H3/t17-,18+,19+,20-,21-,22-,24-/m0/s1
SMILES C[C@]12CCC3(CC1=CC[C@H]4[C@H]5CC[C@](O)([C@@]5(C)C[C@@H](O)[C@@H]24)C6(CO)OCCO6)OCCO3
IUPAC Name (8'S,9'R,10'R,11'R,13'S,14'R,17'S)-17'-[2-(hydroxymethyl)-1,3-dioxolan-2-yl]-10',13'-dimethylspiro[1,3-dioxolane-2,3'-2,4,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene]-11',17'-diol
Molecular Weight (g/mol) 450.57
Formula Weight 450.2618
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.