missing translation for 'onlineSavingsMsg'
Learn More

11Alpha,17Alpha,21-Trihydroxy-pregn-5-ene-3,20-dione 3,20-Diethylene Ketal, TRC

Product Code. 30450680
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450680 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450680 Supplier Toronto Research Chemicals Supplier No. TRCT795245500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11alpha,17alpha,21-Trihydroxy-pregn-5-ene-3,20-dione 3,20-Diethylene Ketal
CAS 102030-55-7
Molecular Formula C25H38O7
Quantity 500 mg
Synonym 11,17,21-Trihydroxy-pregn-5-ene-3,20-dione Cyclic 3,20-Bis(1,2-ethanediyl Acetal)
InChI Formula InChI=1S/C25H38O7/c1-21-7-8-23(29-9-10-30-23)13-16(21)3-4-17-18-5-6-24(28,25(15-26)31-11-12-32-25)22(18,2)14-19(27)20(17)21/h3,17-20,26-28H,4-15H2,1-2H3/t17-,18+,19+,20-,21-,22-,24-/m0/s1
SMILES C[C@]12CCC3(CC1=CC[C@H]4[C@H]5CC[C@](O)([C@@]5(C)C[C@@H](O)[C@@H]24)C6(CO)OCCO6)OCCO3
IUPAC Name (8'S,9'R,10'R,11'R,13'S,14'R,17'S)-17'-[2-(hydroxymethyl)-1,3-dioxolan-2-yl]-10',13'-dimethylspiro[1,3-dioxolane-2,3'-2,4,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene]-11',17'-diol
Molecular Weight (g/mol) 450.57
Formula Weight 450.2618
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.