missing translation for 'onlineSavingsMsg'
Learn More

11a-Hydroxyandrost-4-ene-3,17-dione (>85%), TRC

Product Code. 30397790
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397790 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397790 Supplier Toronto Research Chemicals Supplier No. TRCH80151525MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11Alpha-Hydroxyandrost-4-ene-3,17-dione
CAS 564-33-0
Molecular Formula C19 H26 O3
Quantity 25 mg
Synonym (11α)-11-Hydroxyandrost-4-ene-3,17-dione, 11α-Hydroxy-4-androsten-3,17-dione, 11α-Hydroxyandrost-4-ene-3,17-dione, 11α-Hydroxyandrostendione, 11α-Hydroxyandrostenedione, Androst-4-en-11α-ol-3,17-dione, NSC 9720, 4-Androsten-11alpha-ol-3,17-dione, 4-ANDROSTEN-11a-OL-3,17-DIONE
InChI Formula InChI=1S/C19H26O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h9,13-15,17,21H,3-8,10H2,1-2H3/t13-,14-,15+,17+,18-,19-/m0/s1
SMILES C[C@]12C[C@@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O
IUPAC Name (8S,9S,10R,11R,13S,14S)-11-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-3,17-dione
Molecular Weight (g/mol) 302.4079
Formula Weight 302.1882
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.