missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ 11406 Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters

Product Code. 10673321 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
50 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10673321 50 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10673321 Supplier Sartorius™ Supplier No. 1140650ACN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

11406 Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters have a very uniform pore structure and high non-specific adsorption. Single-packed or dispenser-ready, these individually wrapped filters are ideal for colony counting, particle testing, microscopy, and sterility testing.

  • Product Type: Membrane Filter
  • Material: Cellulose Nitrate
  • Membrane Color/Grid Color: White/Black
  • Pore Size: 0.45 μm
  • Sterility: Sterile
  • For Use With (Application): Bioburden/Microbial Enumeration, Food and Beverage, Environmental/Water Testing
TRUSTED_SUSTAINABILITY

Specifications

Color White
Disposable Single-use
Filter Type Membrane Filter
Format Disc
Material Cellulose Nitrate
Pore Size 0.45 μm
Sterility Sterile
For Use With (Application) Bioburden/Microbial Enumeration, Food and Beverage, Environmental/Water Testing
Grid Color Black
No. per Pack 100 Pk.
Quantity 100 Pk.
Thickness (English) 4.5 to 5.7 mil
Thickness (Metric) 115 to 145 μm
Type Membrane Filter
Diameter (English) 2 in.
Diameter (Metric) 50 mm
Show More Show Less
compliance-icons
Product Identifier
  • CN white/black, sterile, 0.45 micron, 100pk
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P230-Keep wetted with .
  • P243-Take precautionary measures against static discharge.
  • P234-Keep only in original container.
  • P233-Keep container tightly closed.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P372-Explosion risk in case of fire.
  • P375-Fight fire remotely due to the risk of explosion.
  • P370+P380-In case of fire: Evacuate area.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Nitrogen in nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.