Learn More
Thermo Scientific Alfa Aesar Potassium pentachlororhodate(III), Premion™, 99.99% (metals basis), Rh 28.2% min
Brand: Thermo Scientific Alfa Aesar 040409.06
Potassium pentachlororhodate(III), Premion™, 99.99% (metals basis), Rh 28.2% min
Code : NEW
Additional Details : CAS Number : 65980-75-8 Weight : 0.00500kg