Learn More
Thermo Scientific Alfa Aesar Cobalt foil, 2.0mm (0.08 in.) thick, Puratronic™, 99.995% (metals basis)
Brand: Thermo Scientific Alfa Aesar 011449.FI
Cobalt foil, 2.0mm (0.08 in.) thick, Puratronic™, 99.995% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-48-4 Weight : 0.01000kg