missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Copper Manganese Nickel wire, 0.5mm (0.02 in.) dia.

Brand:  Thermo Scientific Alfa Aesar 043190.H9

Copper Manganese Nickel wire, 0.5mm (0.02 in.) dia.

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11354888

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.