Learn More
Thermo Scientific Alfa Aesar Lanthanum foil, 0.3mm (0.012 in.) thick, 99.9% (REO)
Brand: Thermo Scientific Alfa Aesar 010219.FF
Lanthanum foil, 0.3mm (0.012 in.) thick, 99.9% (REO)
Code : NEW
Additional Details : CAS Number : 7439-91-0 Weight : 0.01000kg Transport : UN number : 3208 Chem class : 4.3 Pack group : II