Learn More
KNF Neuberger™ Aluminium/Chromium Diaphragm Pump
Transfer, compress and pump down without contamination. KNF™ Aluminium/Chromium Diaphragm Pumps are double-head, dry-running devices used in a wide range of laboratory applications.
Brand: KNF Neuberger™ 049268/018121
Description
- Available in various versions differing in the materials which contact the media
- Good chemical resistance
- Ideal for sampling, evacuating and compressing applications
- All pump types are based on the same working principle of an oscillating diaphragm inside the pump head offering oil-free, uncontaminated and maintenance-free operation
Specifications
| Diaphragms, Valves, Joints, Screws | |
| 103 mm | |
| 80 w | |
| 15 L/min. | |
| 194 mm | |
| Diaphragm Pump | |
| 230 V |
| 230 V 50 Hz 80 w | |
| Aluminum, Chromium | |
| 4 kg | |
| 50 Hz | |
| 203 mm | |
| 4 bar |