missing translation for 'onlineSavingsMsg'
Learn More

11-(Pentafluorophenoxy)undecyltriethoxysilane, 95%, Thermo Scientific™

Product Code. p-7031837
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428295 250 mg 250mg
15425847 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428295 Supplier Thermo Scientific Alfa Aesar Supplier No. H33057.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197981-13-7
Molecular Formula C23H37F5O4Si
Molecular Weight (g/mol) 500.622
MDL Number MFCD13181914
InChI Key YAAQJDPGHMLLJH-UHFFFAOYSA-N
Synonym 11-pentafluorophenoxyundecyl triethoxysilane, 11-pentafluorophenoxy undecyltriethoxysilane, triethoxy 11-2,3,4,5,6-pentafluorophenoxy undecyl silane
PubChem CID 99738222
IUPAC Name triethoxy-[11-(2,3,4,5,6-pentafluorophenoxy)undecyl]silane
SMILES CCO[Si](CCCCCCCCCCCOC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC

Specifications

Boiling Point 150°C to 155°C (0.04mmHg)
Quantity 250 mg
Formula Weight 500.62
Percent Purity 95%
Refractive Index 1.4340-1.4380
Sensitivity Moisture sensitive
Chemical Name or Material 11-(Pentafluorophenoxy)undecyltriethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.