missing translation for 'onlineSavingsMsg'
Learn More

11-oxo-mogroside V, MedChemExpress

Product Code. 16492529
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492529 1 mg 1mg
16478998 10 mg 10mg
16411428 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492529 Supplier Medchem Express Supplier No. HYN0501

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 11-oxo-mogroside V is a natural sweetener that exhibits strong antioxidant activity. It exhibits significant inhibitory effects on reactive oxygen species (O2-, H2O2 and *OH) with EC50 of 4.79, 16.52, and 146.17 μg/mL, respectively.

11-oxo-mogroside V is a natural sweetener that exhibits strong antioxidant activity. It exhibits significant inhibitory effects on reactive oxygen species (O2-, H2O2 and *OH) with EC50 of 4.79, 16.52, and 146.17 μg/mL, respectively.

Specifications

Chemical Name or Material 11-oxo-mogroside V
CAS 126105-11-1
Color White
Purity Grade Notes Research
Molecular Formula C60H100O29
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (77.80 mM; Need ultrasonic)
SMILES C[C@@]([C@@](CC[C@H](O[C@]([C@@H]([C@@H](O)[C@@H]1O)O)([H])O[C@@H]1CO[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO)C3(C)C)([H])C3=CC4)(C(C[C@@]56C)=O)[C@]4([H])[C@@]5(CC[C@]6([H])[C@H](C)CC[C@H](C(C)(O)C)O[C@H](O[C@H](CO[C@@H]([C@@H]([C@@H](O)[C@@H]7O)O)O[C@@H]7CO)[C@@H](O)[C@@H]8O)[C@@H]8O[C@]([C@@H]([C@@H](O)[C@@H]9O)O)([H])O[C@@H]9CO)C
Molecular Weight (g/mol) 1285.42
Formula Weight 1285.42
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.