missing translation for 'onlineSavingsMsg'
Learn More

11-Hydroxy Dihydro Loratadine, TRC

Product Code. 30397678
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397678 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397678 Supplier Toronto Research Chemicals Supplier No. TRCH94137050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4-[(11RS)-8-Chloro-11-hydroxy-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate
CAS 133284-74-9
Purity Grade Notes HPLC
Molecular Formula C22 H25 Cl N2 O3
Quantity 50 mg
Synonym Ethyl 4-[(11RS)-8-Chloro-11-hydroxy-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate, Loratadine Imp. A (EP)
InChI Formula InChI=1S/C22H25ClN2O3/c1-2-28-21(26)25-12-9-17(10-13-25)22(27)19-8-7-18(23)14-16(19)6-5-15-4-3-11-24-20(15)22/h3-4,7-8,11,14,17,27H,2,5-6,9-10,12-13H2,1H3
SMILES CCOC(=O)N1CCC(CC1)C2(O)c3ccc(Cl)cc3CCc4cccnc24
IUPAC Name ethyl 4-(8-chloro-11-hydroxy-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-yl)piperidine-1-carboxylate
Molecular Weight (g/mol) 400.9
Formula Weight 400.1554
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.