missing translation for 'onlineSavingsMsg'
Learn More

11-Ferrocenyl-1-undecanethiol, TRC

Product Code. 30397038
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397038 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397038 Supplier Toronto Research Chemicals Supplier No. TRCF30763510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Ferrocenyl-1-undecanethiol
CAS 127087-36-9
Molecular Formula C21H32FeS
Quantity 10 mg
Synonym (11-Mercaptoundecyl)ferrocene, 11-Ferrocenyl-1-undecanethiol, 11-Ferrocenylundecanethiol, Ferrocenylundecanethiol, ω-Mercaptoundecylferrocene
InChI Formula InChI=1S/C16H27S.C5H5.Fe/c17-15-11-7-5-3-1-2-4-6-8-12-16-13-9-10-14-16;1-2-4-5-3-1;/h9-10,13-14,17H,1-8,11-12,15H2;1-5H;/q2*-1;+2
SMILES SCCCCCCCCCCC[C-]12[CH]3=[CH]4[CH]5=[CH]1[Fe+2]67894532[CH]=%10[CH]9=[CH]8[CH-]7[CH]%106
IUPAC Name 11-Ferrocenyl-1-undecanethiol
Molecular Weight (g/mol) 372.39
Formula Weight 372.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.