missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Deoxy Corticosterone, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 21-Hydroxyprogesterone |
| CAS | 64-85-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21 H30 O3 |
| Quantity | 100 mg |
| Synonym | Pregn-4-ene-3,20-dione, 21-hydroxy-, 11-Deoxycorticosterone (8CI), 11-Dehydroxycorticosterone, 11-Desoxycorticosterone, 21-Hydroxy-3,20-dioxopregn-4-ene, 21-Hydroxy-δ4-pregnane-3,20-dione, 21-Hydroxy-δ4-pregnene-3,20-dione, 21-Hydroxypregn-4-ene-3,20-dione, 21-Hydroxyprogesterone, 4-Pregnen-21-ol-3,20-dione, Cortexone, Corticosterone, 11-deoxy-, DOC, DOC (steroid), Deoxycorticosterone, Deoxycortone, Desoxycorticosterone, Desoxycortone, Kendall's desoxy compound B, NSC 11319, Reichstein's substance Q, δ4-Pregnene-21-ol-3,20-dione |
| InChI Formula | InChI=1S/C21H30O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h11,15-18,22H,3-10,12H2,1-2H3/t15-,16-,17-,18+,20-,21-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO |
| IUPAC Name | (8S,9S,10R,13S,14S,17S)-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 330.46 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction