missing translation for 'onlineSavingsMsg'
Learn More

11-Cyclopropyl-5,11-dihydro-4-[[(methylsulfonyl)oxy]methyl]-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, TRC

Product Code. 30349765
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30349765 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349765 Supplier Toronto Research Chemicals Supplier No. TRCC9920105MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Cyclopropyl-5,11-dihydro-4-(((methylsulfonyl)oxy)methyl)-6H-dipyrido(3,2-b:2',3'-e)(1,4)diazepin-6-one
CAS 1046462-02-5
Molecular Formula C16H16N4O4S
Quantity 5 mg
Synonym (11-Cyclopropyl-6-oxo-6,11-dihydro-5H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-4-yl)methyl methanesulfonate
InChI Formula InChI=1S/C16H16N4O4S/c1-25(22,23)24-9-10-6-8-18-15-13(10)19-16(21)12-3-2-7-17-14(12)20(15)11-4-5-11/h2-3,6-8,11H,4-5,9H2,1H3,(H,19,21)
SMILES O=S(OCC1=C2C(N(C3CC3)C(N=CC=C4)=C4C(N2)=O)=NC=C1)(C)=O
IUPAC Name (11-cyclopropyl-6-oxo-6,11-dihydro-5H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-4-yl)methyl methanesulfonate
Molecular Weight (g/mol) 360.39
Formula Weight 360.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.