missing translation for 'onlineSavingsMsg'
Learn More

11-(Chloroacetyl)-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one, TRC

Product Code. 30366154
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
Unit Size:
2g
5g
Product Code. Quantity unitSize
30366154 2 g 2g
30359551 10 g -
30351961 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 30366154

Brand: Toronto Research Chemicals TRCC3634052G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28797-48-0
Formula Weight 287.0462
InChI Formula InChI=1S/C14H10ClN3O2/c15-8-12(19)18-11-6-2-1-4-9(11)14(20)17-10-5-3-7-16-13(10)18/h1-7H,8H2,(H,17,20)
IUPAC Name 11-(2-chloroacetyl)-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
Molecular Formula C14H10ClN3O2
Molecular Weight (g/mol) 287.7
SMILES ClCC(=O)N1c2ccccc2C(=O)Nc3cccnc13
Chemical Name or Material 11-(Chloroacetyl)-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one

Specifications

Quantity 2 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.