missing translation for 'onlineSavingsMsg'
Learn More

11-Chloro-1-undecene, 97%

Product Code. 15490268
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490268 5 mL 5mL
15400278 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490268 Supplier Thermo Scientific Alfa Aesar Supplier No. H53449.AA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 872-17-3 | C11H21Cl | 188.74 g/mol

11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-17-3
Molecular Formula C11H21Cl
Molecular Weight (g/mol) 188.74
MDL Number MFCD00671354
InChI Key PSEVKFKRYVAODC-UHFFFAOYSA-N
Synonym 11-chloro-1-undecene, 1-undecene, 11-chloro, 10-undecenyl chloride, undecylenyl chloride, acmc-1bjnc, undec-10-enyl chloride, 11-chloro-1-undecene #
PubChem CID 543805
IUPAC Name 11-chloroundec-1-ene
SMILES ClCCCCCCCCCC=C

Specifications

Density 0.995
Boiling Point 108°C to 109°C (6 mmHg)
Flash Point 104°C (219°F)
Linear Formula Cl(CH2)9CH=CH2
Refractive Index 1.451
Quantity 5 mL
Solubility Information Soluble in water.
Formula Weight 188.74
Percent Purity 97%
Chemical Name or Material 11-Chloro-1-undecene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.