missing translation for 'onlineSavingsMsg'
Learn More

11-Chloro-1-undecene 97%, Thermo Scientific™

Product Code. 10578463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10578463 25mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10578463 Supplier Thermo Scientific Acros Supplier No. 380840250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 872-17-3
CAS Min % 96.0
CAS Max % 100.0
Density 0.9950g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H21Cl
Linear Formula CH2CH(CH2)9Cl
Refractive Index 1.4500 to 1.4520 (20°C, 589nm)
MDL Number MFCD00671354
Quantity 25mL
Synonym 11-chloro-1-undecene, 1-undecene, 11-chloro, 10-undecenyl chloride, undecylenyl chloride, acmc-1bjnc, undec-10-enyl chloride, 11-chloro-1-undecene #
InChI Key PSEVKFKRYVAODC-UHFFFAOYSA-N
SMILES ClCCCCCCCCCC=C
IUPAC Name 11-chloroundec-1-ene
Molecular Weight (g/mol) 188.74
PubChem CID 543805
Formula Weight 188.74
Percent Purity ≥96.0%
Chemical Name or Material 11-Chloro-1-undecene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.