missing translation for 'onlineSavingsMsg'
Learn More

11-Bromoundecanol, 97%, Thermo Scientific Chemicals

Product Code. 11373526
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

11-Bromoundecanol was used in the preparation of 11-mercapto-1-undecanol1 and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Bromoundecanol was used in the preparation of 11-mercapto-1-undecanol1 and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1611-56-9
Molecular Formula C11H23BrO
Molecular Weight (g/mol) 251.21
MDL Number MFCD00004752
InChI Key XFGANBYCJWQYBI-UHFFFAOYSA-N
Synonym 11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a
PubChem CID 74163
IUPAC Name 11-bromoundecan-1-ol
SMILES OCCCCCCCCCCCBr

Specifications

Melting Point 46°C to 49°C
Boiling Point 165°C to 170°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1741180
Solubility Information Insoluble in water.
Formula Weight 251.21
Percent Purity 97%
Chemical Name or Material 11-Bromoundecanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'