missing translation for 'onlineSavingsMsg'
Learn More

11-Bromoundecanoic Acid, 98%

Product Code. 10257273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10257273 1 kg Glass bottle 1kg
10240163 25 g Glass bottle 25g
10747333 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10257273 Supplier Thermo Scientific Acros Supplier No. 423160010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2834-05-1 | C11H21BrO2 | 265.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2834-05-1
Molecular Formula C11H21BrO2
Molecular Weight (g/mol) 265.19
MDL Number MFCD00002732
InChI Key IUDGNRWYNOEIKF-UHFFFAOYSA-N
Synonym undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid
PubChem CID 17812
IUPAC Name 11-bromoundecanoic acid
SMILES C(CCCCCBr)CCCCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 51°C
Color White to Beige
Boiling Point 173°C to 174°C (2.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)10CO2H
Quantity 1 kg
Beilstein 02, II, 315
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols and hydrocarbons
Formula Weight 265.19
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material 11-Bromoundecanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 11-Bromoundecanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.