missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecanol, TRC

Product Code. 30328838
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328838 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328838 Supplier Toronto Research Chemicals Supplier No. TRCB68889010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Bromo-1-undecanol
CAS 1611-56-9
Molecular Formula C11 H23 Br O
Quantity 10 g
Synonym 1-Bromo-11-hydroxyundecane, 1-Bromo-11-undecanol, 11-Bromo-1-hydroxyundecane, 11-Bromoundecan-1-ol, 11-Bromoundecanol, 11-Bromoundecanyl Alcohol, NSC 4029, ω-Bromoundecyl Alcohol
InChI Formula InChI=1S/C11H23BrO/c12-10-8-6-4-2-1-3-5-7-9-11-13/h13H,1-11H2
SMILES OCCCCCCCCCCCBr
IUPAC Name 11-bromoundecan-1-ol
Molecular Weight (g/mol) 251.204
Formula Weight 250.093
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.