missing translation for 'onlineSavingsMsg'
Learn More

11-Aminoundecyltriethoxysilane, 90+%, Thermo Scientific Chemicals

Catalog Number 15456707
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
116821-45-5
C17H39NO3Si
333.588
MFCD08272792
LPWZCJFZJCOBHO-UHFFFAOYSA-N
18944844
11-triethoxysilylundecan-1-amine
CCO[Si](CCCCCCCCCCCN)(OCC)OCC
This item is not returnable. View return policy
116821-45-5
C17H39NO3Si
333.588
MFCD08272792
LPWZCJFZJCOBHO-UHFFFAOYSA-N
18944844
11-triethoxysilylundecan-1-amine
CCO[Si](CCCCCCCCCCCN)(OCC)OCC

Used as primary and secondary intermediate for pharmaceutical and chemical additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical and chemical additives.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Boiling Point 110°C to 115°C (0.02mmHg)
Quantity 250 mg
Linear Formula H2N(CH2)11Si(OCH2CH3)3
Synonym 11-triethoxysilyl-1-undecanamine, 11-aminoundecyltriethoxysilane, 11-triethoxysilyl undecan-1-amine, 1-undecanamine,11-triethoxysilyl, 11-aminoundecyl triethoxysilane, acmc-20mmty, 1-undecanamine, 11-triethoxysilyl
Formula Weight 333.59
Percent Purity 95%
Sensitivity Moisture sensitive
Chemical Name or Material 11-Aminoundecyltriethoxysilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'