missing translation for 'onlineSavingsMsg'
Learn More

11-(Aminooxy)undecyltrimethoxysilane, 95%, Thermo Scientific™

Product Code. 15425797
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425797 250 mg 250mg
15415797 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425797 Supplier Thermo Scientific Alfa Aesar Supplier No. H33007.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870482-12-5
Molecular Formula C14H33NO4Si
Molecular Weight (g/mol) 307.506
MDL Number MFCD11046088
InChI Key SDAAYIFGIWPAIZ-UHFFFAOYSA-N
Synonym 11-o-hydroxylamine undecyltrimethoxysilane, 11-aminooxy undecyl trimethoxysilane, o-11-trimethoxysilyl undecyl hydroxylamine
PubChem CID 45158967
IUPAC Name O-(11-trimethoxysilylundecyl)hydroxylamine
SMILES CO[Si](CCCCCCCCCCCON)(OC)OC

Specifications

Boiling Point 125°C to 130°C (0.08mmHg)
Quantity 250 mg
Linear Formula H2NO(CH2)11Si(OCH3)3
Formula Weight 307.51
Percent Purity 95%
Refractive Index 1.4370-1.4410
Sensitivity Moisture sensitive
Chemical Name or Material 11-(Aminooxy)undecyltrimethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.