missing translation for 'onlineSavingsMsg'
Learn More

11-[4-[2-(2-Ethoxyethoxy)ethyl]piperazin-1-yl]dibenzo[b,f][1,4]thiazepine, Mikromol™

Product Code. 30050553
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050553 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050553 Supplier Mikromol Supplier No. MM1130.510050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

11-[4-[2-(2-Ethoxyethoxy)ethyl]piperazin-1-yl]dibenzo[b,f][1,4]thiazepine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-[4-[2-(2-Ethoxyethoxy)ethyl]piperazin-1-yl]dibenzo[b,f][1,4]thiazepine
CAS 1011758-06-7
Analyte or Component Names 11-[4-[2-(2-Ethoxyethoxy)ethyl]piperazin-1-yl]dibenzo[b,f][1,4]thiazepine
Shipping Condition Room Temperature
Molecular Formula C23 H29 N3 O2 S
Certifications/Compliance ISO 9001
Synonym Dibenzo[b,f][1,4]thiazepine, 11-[4-[2-(2-ethoxyethoxy)ethyl]-1-piperazinyl]-, 11-[4-[2-(2-Ethoxyethoxy)ethyl]-1-piperazinyl]dibenzo[b,f][1,4]thiazepine, 11-[4-[2-(2-Ethoxyethoxy)ethyl]piperazin-1-yl]dibenzo[b,f][1,4]thiazepine
InChI Formula InChI=1S/C23H29N3O2S/c1-2-27-17-18-28-16-15-25-11-13-26(14-12-25)23-19-7-3-5-9-21(19)29-22-10-6-4-8-20(22)24-23/h3-10H,2,11-18H2,1H3
SMILES CCOCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24
IUPAC Name 6-[4-[2-(2-ethoxyethoxy)ethyl]piperazin-1-yl]benzo[b][1,4]benzothiazepine
Molecular Weight (g/mol) 411.56
Quantity 50 mg
Formula Weight 411.198 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.