missing translation for 'onlineSavingsMsg'
Learn More

11-[3-(Dimethylamino)propyl]-6,11-dihydrocibenz[b,e]oxepin-11-ol, TRC

Product Code. 30380112
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380112 250 mg
30387585 100 mg
30372394 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380112 Supplier Toronto Research Chemicals Supplier No. TRCD461765250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4504-88-5
Molecular Formula C19 H23 N O2
Molecular Weight (g/mol) 297.39
Synonym (11RS)-11-[3-(Dimethylamino)-propyl]-6,11-dihydrodibenzo[b,e]oxepin-11-ol, Doxepin Hydrochloride Imp. B (EP), Doxepinol
IUPAC Name 11-[3-(dimethylamino)propyl]-6H-benzo[c][1]benzoxepin-11-ol
SMILES CN(C)CCCC1(O)c2ccccc2COc3ccccc13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H23NO2/c1-20(2)13-7-12-19(21)16-9-4-3-8-15(16)14-22-18-11-6-5-10-17(18)19/h3-6,8-11,21H,7,12-14H2,1-2H3
Formula Weight 297.1729
Percent Purity >95%
Chemical Name or Material (11RS)-11-(3-(Dimethylamino)-propyl)-6,11-dihydrodibenzo(b,e)oxepin-11-ol (Doxepinol)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.